Assay Details
| Gene Symbol: RIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341724.1 | NP_001328533.1 | 11 - 12 | 1575 | 64 | |
| NM_118655.4 | NP_194253.2 | 11 - 12 | 1689 | 64 | ||
| NM_179221.2 | NP_849552.1 | 10 - 11 | 1903 | 64 | ||
| GenBank mRNA | AY072178.1 | - | 10 - 11 | 1690 | 64 | |
More Information
Set Membership: |
|

RIN2
Genomic Map