Assay Details
| Gene Symbol: SHB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118667.3 | NP_194265.2 | 3 - 4 | 1086 | 127 | |
| GenBank mRNA | AY507956.1 | - | 3 - 4 | 905 | 127 | |
More Information
Set Membership: |
|

SHB1
Genomic Map