Assay Details
| Gene Symbol: BETA-OHASE 1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036638.3 | NP_001031715.1 | 2 - 3 | 504 | 88 | |
| NM_001341759.1 | NP_001320065.1 | 2 - 3 | 618 | 88 | ||
| NM_118702.4 | NP_194300.1 | 2 - 3 | 618 | 88 | ||
| GenBank mRNA | AF125576.1 | - | 2 - 3 | 504 | 88 | |
| AF370220.1 | - | 2 - 3 | 498 | 88 | ||
| AY113923.1 | - | 2 - 3 | 435 | 88 | ||
More Information
Set Membership: |
|

BETA-OHASE 1
Genomic Map