Assay Details
| Gene Symbol: AT4G25830 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118715.4 | NP_567729.1 | 1 - 2 | 477 | 117 | |
| GenBank mRNA | AK175371.1 | - | 1 - 2 | 350 | 117 | |
| AK175732.1 | - | 1 - 2 | 235 | 117 | ||
| AK176002.1 | - | 1 - 2 | 229 | 117 | ||
| AK176011.1 | - | 1 - 2 | 229 | 117 | ||
| AK176089.1 | - | 1 - 2 | 241 | 117 | ||
| AK229626.1 | - | 1 - 2 | 243 | 117 | ||
| AY088797.1 | - | 1 - 2 | 243 | 117 | ||
More Information
Set Membership: |
|

AT4G25830
Genomic Map