Assay Details
Gene Symbol: ABI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118741.3 | NP_194338.1 | 2 - 3 | 1383 | 68 | |
GenBank mRNA | AK226529.1 | - | 2 - 3 | 982 | 68 | |
AY142623.1 | - | 2 - 3 | 835 | 68 | ||
U12856.1 | - | 2 - 3 | 1266 | 68 | ||
X77116.1 | - | 1266 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
