Assay Details
| Gene Symbol: RPS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118742.2 | NP_194339.1 | 1 | 3057 | 129 | |
| GenBank mRNA | AK117214.1 | - | 1 | 2820 | 129 | |
| U12860.1 | - | 1 | 2831 | 129 | ||
More Information
Set Membership: |
|

RPS2
Genomic Map