Assay Details
Gene Symbol: BGAL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341777.1 | NP_001329560.1 | 13 - 14 | 1693 | 72 | |
NM_001341780.1 | NP_001329558.1 | 13 - 14 | 1642 | 72 | ||
NM_001341782.1 | NP_001329555.1 | 13 - 14 | 1641 | 72 | ||
NM_118747.3 | NP_194344.2 | 13 - 14 | 1693 | 72 | ||
NM_179222.2 | NP_849553.1 | 13 - 14 | 1928 | 72 | ||
GenBank mRNA | AJ270308.1 | - | 13 - 14 | 1538 | 72 | |
AY099612.1 | - | 13 - 14 | 1635 | 72 | ||
BT000267.1 | - | 13 - 14 | 1532 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
