Assay Details
Gene Symbol: BGAL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341777.1 | NP_001329560.1 | 14 - 15 | 1802 | 138 | |
NM_001341780.1 | NP_001329558.1 | 14 - 15 | 1751 | 138 | ||
NM_001341781.1 | NP_001329559.1 | 13 - 14 | 1826 | 138 | ||
NM_001341783.1 | NP_001329556.1 | 13 - 14 | 1825 | 138 | ||
NM_118747.3 | NP_194344.2 | 14 - 15 | 1802 | 138 | ||
NM_179222.2 | NP_849553.1 | 14 - 15 | 2037 | 138 | ||
GenBank mRNA | AJ270308.1 | - | 14 - 15 | 1647 | 138 | |
AY099612.1 | - | 14 - 15 | 1744 | 138 | ||
BT000267.1 | - | 14 - 15 | 1641 | 138 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
