Assay Details
| Gene Symbol: FBA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036644.2 | NP_001031721.1 | 3 - 4 | 469 | 70 | |
| NM_001341809.1 | NP_001328708.1 | 2 - 3 | 1099 | 70 | ||
| NM_118786.3 | NP_194383.1 | 3 - 4 | 412 | 70 | ||
| GenBank mRNA | AY087376.1 | - | 2 - 3 | 347 | 70 | |
| BT002006.1 | - | 2 - 3 | 347 | 70 | ||
| BT008844.1 | - | 2 - 3 | 287 | 70 | ||
More Information
Set Membership: |
|

FBA5
Genomic Map