Assay Details
Gene Symbol: AT4G26630 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036647.3 | NP_001031724.1 | 7 - 8 | 1594 | 67 | |
NM_118797.5 | NP_194393.3 | 7 - 8 | 1585 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
