Assay Details
| Gene Symbol: TOR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118840.5 | NP_194436.2 | 2 - 3 | 1094 | 69 | |
| GenBank mRNA | AB176521.1 | - | 2 - 3 | 914 | 69 | |
| AJ249836.2 | - | 2 - 3 | 993 | 69 | ||
More Information
Set Membership: |
|

TOR1
Genomic Map