Assay Details
Gene Symbol: COBL11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118845.3 | NP_567766.1 | 2 - 3 | 782 | 60 | |
GenBank mRNA | AK175703.1 | - | 2 - 3 | 748 | 60 | |
AK176063.1 | - | 2 - 3 | 748 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
