Assay Details
| Gene Symbol: UBP13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202935.1 | NP_001189864.1 | 14 - 15 | 1977 | 70 | |
| NM_001337961.1 | NP_001319526.1 | 14 - 15 | 1977 | 70 | ||
| NM_001337962.1 | NP_001326292.1 | 14 - 15 | 1980 | 70 | ||
| NM_112024.6 | NP_187797.3 | 14 - 15 | 1906 | 70 | ||
| GenBank mRNA | BT002760.1 | - | 14 - 15 | 1707 | 70 | |
More Information
Set Membership: |
|

UBP13
Genomic Map