Assay Details
| Gene Symbol: emb2738 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084670.1 | NP_001078139.1 | 9 - 10 | 1664 | 81 | |
| NM_112042.4 | NP_187815.2 | 9 - 10 | 1713 | 81 | ||
| GenBank mRNA | BT046184.1 | - | 9 - 10 | 1605 | 81 | |
More Information
Set Membership: |
|

emb2738
Genomic Map