Assay Details
Gene Symbol: Nek7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202938.1 | NP_001189867.1 | 7 - 8 | 921 | 74 | |
NM_001337978.1 | NP_001327416.1 | 5 - 6 | 968 | 74 | ||
NM_001337979.1 | NP_001327417.1 | 6 - 7 | 1182 | 74 | ||
NM_112055.5 | NP_187827.1 | 6 - 7 | 822 | 74 | ||
GenBank mRNA | BT015419.1 | - | 6 - 7 | 441 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
