Assay Details
Gene Symbol: RBR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202939.1 | NP_001189868.1 | 14 - 15 | 2799 | 107 | |
NM_112064.5 | NP_566417.3 | 14 - 15 | 2988 | 107 | ||
GenBank mRNA | AF245395.1 | - | 14 - 15 | 2706 | 107 | |
AY150438.1 | - | 13 - 14 | 2502 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
