Assay Details
| Gene Symbol: NUDT16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112095.4 | NP_566428.1 | 1 - 2 | 416 | 128 | |
| GenBank mRNA | AF325001.2 | - | 1 - 2 | 302 | 128 | |
| AF372914.1 | - | 1 - 2 | 277 | 128 | ||
| BT002659.1 | - | 1 - 2 | 76 | 128 | ||
More Information
Set Membership: |
|

NUDT16
Genomic Map