Assay Details
Gene Symbol: GrxC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341931.1 | NP_001328515.1 | 1 - 2 | 347 | 129 | |
NM_119017.5 | NP_194602.2 | 1 - 2 | 400 | 129 | ||
GenBank mRNA | AK118672.1 | - | 1 - 2 | 347 | 129 | |
BT003703.1 | - | 1 - 2 | 269 | 129 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
