Assay Details
Gene Symbol: ROP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119039.3 | NP_194624.1 | 2 - 3 | 556 | 121 | |
GenBank mRNA | AF079484.1 | - | 2 - 3 | 353 | 121 | |
BT025271.1 | - | 2 - 3 | 189 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
