Assay Details
Gene Symbol: ROP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119039.3 | NP_194624.1 | 3 - 4 | 669 | 60 | |
GenBank mRNA | AF079484.1 | - | 3 - 4 | 466 | 60 | |
BT025271.1 | - | 3 - 4 | 302 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
