Assay Details
Gene Symbol: ATR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342001.1 | NP_001328167.1 | 2 - 3 | 628 | 149 | |
NM_119167.4 | NP_194750.1 | 2 - 3 | 598 | 149 | ||
NM_179141.2 | NP_849472.2 | 2 - 3 | 559 | 149 | ||
GenBank mRNA | AF325101.1 | - | 2 - 3 | 423 | 149 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
