Assay Details
| Gene Symbol: AT4G30390 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119185.2 | NP_194768.1 | 1 - 2 | 398 | 91 | |
| GenBank mRNA | AK176331.1 | - | 1 - 2 | 264 | 91 | |
| AY085119.1 | - | 1 - 2 | 163 | 91 | ||
| BT024536.1 | - | 1 - 2 | 119 | 91 | ||
More Information
Set Membership: |
|

AT4G30390
Genomic Map