Assay Details
Gene Symbol: BRS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119207.3 | NP_194790.1 | 8 - 9 | 1343 | 65 | |
GenBank mRNA | BT002334.1 | - | 8 - 9 | 1253 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
