Assay Details
Gene Symbol: PBA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036682.1 | NP_001031759.1 | 3 - 4 | 397 | 57 | |
NM_001203950.1 | NP_001190879.1 | 4 - 5 | 289 | 57 | ||
NM_119279.5 | NP_194858.1 | 3 - 4 | 416 | 57 | ||
GenBank mRNA | AF043529.1 | - | 3 - 4 | 385 | 57 | |
AY052354.1 | - | 3 - 4 | 397 | 57 | ||
AY058240.1 | - | 3 - 4 | 233 | 57 | ||
Y13694.1 | - | 304 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
