Assay Details
Gene Symbol: PBA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203950.1 | NP_001190879.1 | 5 - 6 | 362 | 88 | |
NM_119279.5 | NP_194858.1 | 4 - 5 | 489 | 88 | ||
GenBank mRNA | AF043529.1 | - | 4 - 5 | 458 | 88 | |
AY052354.1 | - | 4 - 5 | 470 | 88 | ||
AY058240.1 | - | 4 - 5 | 306 | 88 | ||
Y13694.1 | - | 377 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
