Assay Details
Gene Symbol: PBA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036682.1 | NP_001031759.1 | 6 - 7 | 660 | 79 | |
NM_001203950.1 | NP_001190879.1 | 7 - 8 | 549 | 79 | ||
NM_119279.5 | NP_194858.1 | 6 - 7 | 676 | 79 | ||
GenBank mRNA | AF043529.1 | - | 6 - 7 | 645 | 79 | |
AY052354.1 | - | 6 - 7 | 657 | 79 | ||
AY058240.1 | - | 6 - 7 | 493 | 79 | ||
Y13694.1 | - | 564 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
