Assay Details
| Gene Symbol: CYP83B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119299.3 | NP_194878.1 | 1 - 2 | 1022 | 60 | |
| GenBank mRNA | AY085195.1 | - | 1 - 2 | 928 | 60 | |
| BT002333.1 | - | 1 - 2 | 889 | 60 | ||
| D78598.1 | - | 1 - 2 | 904 | 60 | ||
More Information
Set Membership: |
|

CYP83B1
Genomic Map