Assay Details
| Gene Symbol: ATM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085286.2 | NP_001078755.1 | 17 - 18 | 2603 | 81 | |
| NM_124808.4 | NP_568806.3 | 16 - 17 | 1727 | 81 | ||
More Information
Set Membership: |
|

ATM2
Genomic Map