Assay Details
| Gene Symbol: ANK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124842.4 | NP_200272.2 | 2 - 3 | 743 | 65 | |
| GenBank mRNA | AK117622.1 | - | 2 - 3 | 617 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ANK| Gene Symbol: ANK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124842.4 | NP_200272.2 | 2 - 3 | 743 | 65 | |
| GenBank mRNA | AK117622.1 | - | 2 - 3 | 617 | 65 | |
Set Membership: |
|