Assay Details
Gene Symbol: FTSZ1-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124910.6 | NP_200339.1 | 2 - 3 | 518 | 70 | |
GenBank mRNA | AY034992.1 | - | 2 - 3 | 441 | 70 | |
AY113896.1 | - | 2 - 3 | 396 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
