Assay Details
Gene Symbol: TOP1ALPHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203614.1 | NP_001190543.1 | 15 - 16 | 2819 | 90 | |
NM_001203615.1 | NP_001190544.1 | 9 - 10 | 1756 | 90 | ||
NM_124912.5 | NP_200341.1 | 15 - 16 | 2831 | 90 | ||
GenBank mRNA | X57544.1 | - | 2756 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
