Assay Details
Gene Symbol: TOP1ALPHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203614.1 | NP_001190543.1 | 6 - 7 | 1799 | 78 | |
NM_124912.5 | NP_200341.1 | 6 - 7 | 1811 | 78 | ||
GenBank mRNA | X57544.1 | - | 1736 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
