Assay Details
Gene Symbol: TOP1BETA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124913.4 | NP_200342.1 | 7 - 8 | 1957 | 112 | |
GenBank mRNA | AK226832.1 | - | 7 - 8 | 1873 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
