Assay Details
| Gene Symbol: AT5G55640 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124946.4 | NP_568828.1 | 1 - 2 | 293 | 75 | |
| GenBank mRNA | AF370556.1 | - | 1 - 2 | 216 | 75 | |
| BT006537.1 | - | 1 - 2 | 61 | 75 | ||
More Information
Set Membership: |
|

AT5G55640
Genomic Map