Assay Details
Gene Symbol: AT5G55640 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124946.4 | NP_568828.1 | 2 - 3 | 391 | 121 | |
GenBank mRNA | AF370556.1 | - | 2 - 3 | 314 | 121 | |
BT006537.1 | - | 2 - 3 | 159 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
