Assay Details
| Gene Symbol: CBL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345176.1 | NP_001332655.1 | 5 - 6 | 793 | 103 | |
| NM_124981.4 | NP_200410.1 | 5 - 6 | 793 | 103 | ||
| GenBank mRNA | AF076252.1 | - | 4 - 5 | 331 | 103 | |
| AY052304.1 | - | 4 - 5 | 376 | 103 | ||
| AY139808.1 | - | 3 - 4 | 267 | 103 | ||
More Information
Set Membership: |
|

CBL2
Genomic Map