Assay Details
Gene Symbol: COX15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124991.4 | NP_200420.1 | 1 - 2 | 351 | 70 | |
GenBank mRNA | AK117496.1 | - | 1 - 2 | 226 | 70 | |
BT004976.1 | - | 1 - 2 | 188 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
