Assay Details
| Gene Symbol: CRK6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179095.1 | NP_849426.1 | 5 - 6 | 1569 | 82 | |
| GenBank mRNA | AK118493.1 | - | 5 - 6 | 1569 | 82 | |
More Information
Set Membership: |
|

CRK6
Genomic Map