Assay Details
| Gene Symbol: NUDT10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036636.2 | NP_001031713.1 | 5 - 6 | 774 | 106 | |
| NM_001341732.1 | NP_001329739.1 | 6 - 7 | 672 | 106 | ||
| NM_001341735.1 | NP_001329740.1 | 4 - 5 | 883 | 106 | ||
| NM_001341736.1 | NP_001329746.1 | 4 - 5 | 1062 | 106 | ||
| NM_001341737.1 | NP_001329743.1 | 4 - 5 | 1047 | 106 | ||
| NM_001341738.1 | NP_001329742.1 | 5 - 6 | 968 | 106 | ||
| NM_001341739.1 | NP_001329745.1 | 5 - 6 | 753 | 106 | ||
| NM_001341740.1 | NP_001329744.1 | 6 - 7 | 672 | 106 | ||
| NM_001341741.1 | NP_001329748.1 | 5 - 6 | 753 | 106 | ||
| NM_001341742.1 | NP_001329747.1 | 6 - 7 | 657 | 106 | ||
| NM_179112.3 | NP_849443.2 | 6 - 7 | 657 | 106 | ||
| GenBank mRNA | BT010765.1 | - | 6 - 7 | 621 | 106 | |
| BT010987.1 | - | 6 - 7 | 529 | 106 | ||
More Information
Set Membership: |
|

NUDT10
Genomic Map