Assay Details
| Gene Symbol: CBL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342191.1 | NP_001320119.1 | 4 - 5 | 404 | 74 | |
| NM_001342192.1 | NP_001329201.1 | 4 - 5 | 430 | 74 | ||
| NM_001342193.1 | NP_001329200.1 | 4 - 5 | 430 | 74 | ||
| NM_001342194.1 | NP_001329199.1 | 4 - 5 | 404 | 74 | ||
| NM_119454.2 | NP_195026.1 | 4 - 5 | 358 | 74 | ||
| NM_179154.2 | NP_849485.1 | 4 - 5 | 430 | 74 | ||
| GenBank mRNA | AF490607.1 | - | 4 - 5 | 358 | 74 | |
| AF513507.2 | - | 4 - 5 | 328 | 74 | ||
| AK117439.1 | - | 4 - 5 | 395 | 74 | ||
| BT024464.1 | - | 4 - 5 | 328 | 74 | ||
More Information
Set Membership: |
|

CBL10
Genomic Map