Assay Details
| Gene Symbol: CBL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342192.1 | NP_001329201.1 | 5 - 6 | 548 | 77 | |
| NM_001342193.1 | NP_001329200.1 | 5 - 6 | 548 | 77 | ||
| NM_001342194.1 | NP_001329199.1 | 5 - 6 | 522 | 77 | ||
| NM_119454.2 | NP_195026.1 | 5 - 6 | 476 | 77 | ||
| NM_179154.2 | NP_849485.1 | 5 - 6 | 548 | 77 | ||
| GenBank mRNA | AF490607.1 | - | 5 - 6 | 476 | 77 | |
| AF513507.2 | - | 5 - 6 | 446 | 77 | ||
| AK117439.1 | - | 5 - 6 | 513 | 77 | ||
| BT024464.1 | - | 5 - 6 | 446 | 77 | ||
More Information
Set Membership: |
|

CBL10
Genomic Map