Assay Details
| Gene Symbol: CBL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119454.2 | NP_195026.1 | 7 - 8 | 605 | 164 | |
| NM_179154.2 | NP_849485.1 | 7 - 8 | 677 | 164 | ||
| GenBank mRNA | AF490607.1 | - | 7 - 8 | 605 | 164 | |
| AF513507.2 | - | 7 - 8 | 575 | 164 | ||
| AK117439.1 | - | 7 - 8 | 642 | 164 | ||
| BT024464.1 | - | 7 - 8 | 575 | 164 | ||
More Information
Set Membership: |
|

CBL10
Genomic Map