Assay Details
| Gene Symbol: CBL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342191.1 | NP_001320119.1 | 6 - 7 | 936 | 159 | |
| NM_001342192.1 | NP_001329201.1 | 7 - 8 | 890 | 159 | ||
| NM_119454.2 | NP_195026.1 | 8 - 9 | 718 | 159 | ||
| NM_179154.2 | NP_849485.1 | 8 - 9 | 790 | 159 | ||
| GenBank mRNA | AK117439.1 | - | 8 - 9 | 755 | 159 | |
More Information
Set Membership: |
|

CBL10
Genomic Map