Assay Details
| Gene Symbol: MCA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342380.1 | NP_001329715.1 | 3 - 4 | 838 | 68 | |
| NM_001342381.1 | NP_001329716.1 | 3 - 4 | 726 | 68 | ||
| NM_119759.4 | NP_195317.2 | 3 - 4 | 1066 | 68 | ||
| NM_179172.3 | NP_849503.1 | 3 - 4 | 760 | 68 | ||
| NM_179173.4 | NP_849504.2 | 3 - 4 | 987 | 68 | ||
| GenBank mRNA | AB196960.1 | - | 3 - 4 | 794 | 68 | |
| AK316845.1 | - | 3 - 4 | 761 | 68 | ||
| AY136293.1 | - | 3 - 4 | 818 | 68 | ||
| BT003430.1 | - | 1 - 2 | 410 | 68 | ||
More Information
Set Membership: |
|

MCA1
Genomic Map