Assay Details
| Gene Symbol: MCA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342380.1 | NP_001329715.1 | 4 - 5 | 1253 | 111 | |
| NM_001342381.1 | NP_001329716.1 | 4 - 5 | 1141 | 111 | ||
| NM_119759.4 | NP_195317.2 | 4 - 5 | 1481 | 111 | ||
| NM_179172.3 | NP_849503.1 | 4 - 5 | 1175 | 111 | ||
| NM_179173.4 | NP_849504.2 | 4 - 5 | 1402 | 111 | ||
| GenBank mRNA | AB196960.1 | - | 4 - 5 | 1209 | 111 | |
| AK316845.1 | - | 4 - 5 | 1176 | 111 | ||
| AY136293.1 | - | 4 - 5 | 1233 | 111 | ||
| BT003430.1 | - | 2 - 3 | 825 | 111 | ||
More Information
Set Membership: |
|

MCA1
Genomic Map