Assay Details
Gene Symbol: BGAL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119799.4 | NP_568001.1 | 13 - 14 | 1876 | 67 | |
NM_179175.2 | NP_849506.1 | 13 - 14 | 1671 | 67 | ||
GenBank mRNA | AJ270299.1 | - | 13 - 14 | 1593 | 67 | |
AY056285.1 | - | 13 - 14 | 1652 | 67 | ||
AY091432.1 | - | 13 - 14 | 1553 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
