Assay Details
Gene Symbol: BGAL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119799.4 | NP_568001.1 | 2 - 3 | 602 | 105 | |
NM_179175.2 | NP_849506.1 | 2 - 3 | 397 | 105 | ||
GenBank mRNA | AJ270299.1 | - | 2 - 3 | 319 | 105 | |
AY056285.1 | - | 2 - 3 | 378 | 105 | ||
AY091432.1 | - | 2 - 3 | 279 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
