Assay Details
| Gene Symbol: BEE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119817.3 | NP_195372.1 | 4 - 5 | 935 | 66 | |
| NM_179177.2 | NP_849508.1 | 4 - 5 | 929 | 66 | ||
| GenBank mRNA | AF370231.1 | - | 4 - 5 | 875 | 66 | |
| AY059122.1 | - | 4 - 5 | 819 | 66 | ||
More Information
Set Membership: |
|

BEE2
Genomic Map