Assay Details
Gene Symbol: PIF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331836.1 | NP_001318964.1 | 5 - 6 | 1478 | 69 | |
NM_001331837.1 | NP_001318965.1 | 5 - 6 | 1538 | 69 | ||
NM_001331838.1 | NP_001322700.1 | 4 - 5 | 1902 | 69 | ||
NM_001331839.1 | NP_001322701.1 | 3 - 4 | 1875 | 69 | ||
NM_100824.3 | NP_172424.1 | 4 - 5 | 1451 | 69 | ||
NM_179295.3 | NP_849626.1 | 4 - 5 | 1511 | 69 | ||
GenBank mRNA | AF100166.1 | - | 4 - 5 | 1143 | 69 | |
AF251693.1 | - | 3 - 4 | 1060 | 69 | ||
AK117255.1 | - | 4 - 5 | 1196 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
