Assay Details
Gene Symbol: PIF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331836.1 | NP_001318964.1 | 7 - 8 | 1602 | 69 | |
NM_001331837.1 | NP_001318965.1 | 7 - 8 | 1662 | 69 | ||
NM_001331838.1 | NP_001322700.1 | 6 - 7 | 2026 | 69 | ||
NM_001331839.1 | NP_001322701.1 | 5 - 6 | 1999 | 69 | ||
NM_100824.3 | NP_172424.1 | 6 - 7 | 1575 | 69 | ||
NM_179295.3 | NP_849626.1 | 6 - 7 | 1635 | 69 | ||
GenBank mRNA | AF100166.1 | - | 6 - 7 | 1267 | 69 | |
AF251693.1 | - | 5 - 6 | 1184 | 69 | ||
AK117255.1 | - | 6 - 7 | 1320 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
